Thiophene-2-carbaldehyde

Warning

Synonyms: 2-Thiophenecarboxaldehyde, Thiophene-2-carboxaldehyde

T32409-25G T32409-100G
220226-569EA 34.46 USD
220226-569 220226-568
Thiophene-2-carbaldehyde
Thiophene-2-carbaldehyde
Formula: C₅H₄OS
MW: 112.15 g/mol
Boiling Pt: 198 °C (1013 hPa)
Melting Pt: ? 10 °C
Density: 1.2264 g/cm³ (20 °C)
Flash Pt: 78 °C
MDL Number: MFCD00005429
CAS Number: 98-03-3
EINECS: 202-629-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR