Thioanisole ≥99%, ReagentPlus®

Danger

Synonyms: Methyl phenyl sulphide, Phenyl methyl sulphide, (Methylthio)benzene

T28002-100G T28002-25G T28002-500G
220226-467EA 48.46 USD
220226-467 220226-468 220226-469
Thioanisole ≥99%, ReagentPlus®
Thioanisole
Formula: C₇H₈S
MW: 124.21 g/mol
Boiling Pt: 188 °C (1013 hPa)
Melting Pt: –15 °C
Density: 1.05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008559
CAS Number: 100-68-5
EINECS: 202-878-2
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR