o-Terphenyl ≥99%

Warning

Synonyms: 2-Phenylbiphenyl, 1,2-Terphenyl, 2-Phenyl-1,1'-biphenyl, 1,2-Diphenylbenzene, 1,1:2,1-Terphenyl

T2800-25G T2800-100G
220226-466EA 119.54 USD
220226-466 220226-465
o-Terphenyl ≥99%
o-Terphenyl
Formula: C₁₈H₁₄
MW: 230.31 g/mol
Boiling Pt: 338 °C (1013 hPa)
Melting Pt: 54…56 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 163 °C
Storage Temperature: Ambient
MDL Number: MFCD00003055
CAS Number: 84-15-1
EINECS: 201-517-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR