L(-)-Tryptophan, Reagent Grade

Synonyms: L-Tryptophan, Tryptophan, L-α-Amino-3-indolepropionic acid, (S)-2-Amino-3-(3-indolyl)propionic acid, H-Trp-OH

T0254-100G T0254-25G T0254-1KG T0254-1G T0254-5G T0254-500G
220225-575EA 180.92 USD
220225-575 220225-578 220225-577 220225-576 220225-580 220225-579
L(-)-Tryptophan, Reagent Grade
L(-)-Tryptophan
Formula: C₁₁H₁₂N₂O₂
MW: 204.23 g/mol
Melting Pt: 290 °C
Density: 1.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064340
CAS Number: 73-22-3
EINECS: 200-795-6
Merck Index: 12,09929

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR