Sarcosine

Synonyms: H-Sar-OH, N-Methylglycine, Methylaminoacetic acid

S7672-100G S7672-10MG S7672-25G
220210-240EA 487.38 USD
220210-240 220210-241 220210-242
Sarcosine
Sarcosine
Formula: C₃H₇NO₂
MW: 89.09 g/mol
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 63 °C
Density: 1.301 g/cm³ (20 °C)
MDL Number: MFCD00004279
CAS Number: 107-97-1
EINECS: 203-538-6
Merck Index: 12,08519

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR