Disodium metasilicate nonahydrate

Danger

Synonyms: Sodium metasilicate nonahydrate

S4392-1KG S4392-250G
220209-372EA 296.92 USD
220209-372 220209-373
Disodium metasilicate nonahydrate
Disodium metasilicate nonahydrate
Formula: Na₂SiO₃·9H₂O
MW: 284.2 g/mol
Melting Pt: 40 °C
Density: 2.62 g/cm³ (21 °C)
CAS Number: 13517-24-3
EINECS: 229-912-9
UN: 3253
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR