Spermidine

Synonyms: N1-(3-Aminopropyl)butane-1,4-diamine, N-(3-Aminopropyl)-1,4-diaminobutane, 4-Azaoctamethylenediamine

S2626-25G S2626-1G S2626-5G
220209-056EA 914.17 USD
220209-056 220209-055 220209-057
Spermidine
Spermidine
Formula: C₇H₁₉N₃
MW: 145.25 g/mol
Boiling Pt: 246 °C (1013 hPa)
Melting Pt: 20…25 °C
Density: 0.925 g/cm³ (20 °C)
MDL Number: MFCD00008229
CAS Number: 124-20-9
EINECS: 204-689-0
Merck Index: 12,08893

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR