Rubidium chloride ≥99.0%, ReagentPlus®

Synonyms: Rubidiumchloride

R2252-10G R2252-100G R2252-5G R2252-50G
220207-289EA 115.08 USD
220207-289 220207-288 220207-291 220207-290
Rubidium chloride ≥99.0%, ReagentPlus®
Rubidium chloride
Formula: RbCl
MW: 120.92 g/mol
Boiling Pt: 1390 °C (1013 hPa)
Melting Pt: 715 °C
Density: 2.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011187
CAS Number: 7791-11-9
EINECS: 232-240-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR