Rubrene

Synonyms: 5,6,11,12-Tetraphenylnaphthacene, 5,6,11,12-Tetraphenyltetracene

R2206-100MG R2206-1G
220207-285EA 60.02 USD
220207-285 220207-286
Rubrene
Rubrene
Formula: C₄₂H₂₈
MW: 532.68 g/mol
MDL Number: MFCD00003703
CAS Number: 517-51-1
EINECS: 208-242-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR