Potassium hexacyanoferrate(II) trihydrate ≥99%, ReagentPlus®

Synonyms: Potassiumhexacyanoferrate(II) trihydrate, Tetrapotassium hexacyanoferrate trihydrate, Potassium ferrocyanide trihydrate

P9387-100G P9387-500G P9387-2.5KG
220205-859EA 26.71 USD
220205-859 220205-861 220205-860
Potassium hexacyanoferrate(II) trihydrate ≥99%, ReagentPlus®
Potassium hexacyanoferrate(II) trihydrate
Formula: K₄Fe(CN)₆·3H₂O
MW: 422.39 g/mol
Melting Pt: 70 °C
Density: 1.889 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167023
CAS Number: 14459-95-1
EINECS: 237-722-2
Merck Index: 12,07793

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR