Pyridazine

P57204-5G P57204-50G
220204-396EA 76.14 USD
220204-396 220204-395
Pyridazine
Pyridazine
Formula: C₄H₄N₂
MW: 80.09 g/mol
Boiling Pt: 208 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1.103 g/cm³ (20 °C)
MDL Number: MFCD00006463
CAS Number: 289-80-5
EINECS: 206-025-5
Merck Index: 12,08152

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR