Tripotassium orthophosphate ≥98%, Reagent Grade

Danger

Synonyms: Potassium phosphate tribasic, Potassium phosphate

P5629-500G P5629-25G P5629-10KG P5629-1KG
220204-238EA 58.15 USD
220204-238 220204-237 220204-235 220204-236
Tripotassium orthophosphate ≥98%, Reagent Grade
Tripotassium orthophosphate
Formula: K₃O₄P
MW: 212.27 g/mol
Melting Pt: 1340 °C
Density: 2.564 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036295
CAS Number: 7778-53-2
EINECS: 231-907-1
Merck Index: 12,07830

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR