Pyrazine-2,3-dicarboxylic acid

P56208-100G P56208-25G
220204-230EA 159.38 USD
220204-230 220204-231
Pyrazine-2,3-dicarboxylic acid
Pyrazine-2,3-dicarboxylic acid
Formula: C₆H₄N₂O₄
MW: 168.11 g/mol
MDL Number: MFCD00006131
CAS Number: 89-01-0
EINECS: 201-875-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR