L-Phenylalanine 98.5-101.0%

Synonyms: H-L-Phe-OH, Phenylalanine, H-Phe-OH, L-α-Phenylalanine, (S)-(-)-Phenylalanine, 3-Phenyl-L-alanine, 3-Phenyl-L(-)-alanine

P5482-100G P5482-25G P5482-10MG P5482-1KG
220204-148EA 201.85 USD
220204-148 220204-151 220204-149 220204-150
L-Phenylalanine 98.5-101.0%
L-Phenylalanine
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 173 °C
Density: 0.754 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064227
CAS Number: 63-91-2
EINECS: 200-568-1
Merck Index: 14,07271

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR