Methyl palmitate

Synonyms: Hexadecylic acid methyl ester, Methyl hexadecanoate, Palmitic acid methyl ester, Hexadecanoic acid methyl ester, Palmitic Acid Methyl Ester

P5177-1G P5177-25G P5177-5G
220203-615EA 59.57 USD
220203-615 220203-616 220203-617
Methyl palmitate
Methyl palmitate
Formula: C₁₇H₃₄O₂
MW: 270.46 g/mol
Boiling Pt: 417 °C (1013 hPa)
Melting Pt: 29.9 °C
Density: 0.8804 g/cm³ (20 °C)
MDL Number: MFCD00008994
CAS Number: 112-39-0
EINECS: 203-966-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR