(±)-Pipecolinic acid

Synonyms: (±)-2-Piperidinecarboxylic acid, DL-Pipecolinic acid, H-DL-Homopro-OH, Pipecolinic Acid, Piperidine-2-carboxylic acid, Pipecolinic acid

P45850-25G P45850-100G
220203-837EA 107.69 USD
220203-837 220203-836
(±)-Pipecolinic acid
(±)-Pipecolinic acid
Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
MDL Number: MFCD00064347
CAS Number: 535-75-1
EINECS: 208-616-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR