Pimelic acid ≥98%

Warning

Synonyms: Heptanedioic acid, 1,5-Pentanedicarboxylic acid, 1,7-Heptanedioic acid

P45001-100G P45001-25G
220203-788EA 227.23 USD
220203-788 220203-789
Pimelic acid ≥98%
Pimelic acid
Formula: C₇H₁₂O₄
MW: 160.17 g/mol
Boiling Pt: 354 °C (1013 hPa)
Melting Pt: 101…103 °C
Density: 1.329 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004425
CAS Number: 111-16-0
EINECS: 203-840-8
Merck Index: 12,07585

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR