Potassium chloride 99.0-100.5% ACS
Synonyms:
Potassiumchloride
P3911-25G
P3911-2.5KG
P3911-4X2.5KG
220201-975EA
58.46
USD
220201-975
220201-974
220201-972
220201-973
220203-496
220203-497
220201-976
Potassium chloride 99.0-100.5% ACS
Potassium chloride
Formula:
KCl MW: 74.55 g/mol Boiling Pt: 1420 °C (1013 hPa) Melting Pt: 773 °C Density: 1.98 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011360 CAS Number: 7447-40-7 EINECS: 231-211-8 Merck Index: 14,07621 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...