(±)-1-Phenyl-1,2-ethanediol ≥97%

Synonyms: (±)-α,β-Dihydroxyethylbenzene, (±)-Phenylethane-1,2-diol, 1-Phenylethane-1,2-diol

P24055-5G P24055-100G
220202-984EA 56.75 USD
220202-984 220202-983
(±)-1-Phenyl-1,2-ethanediol ≥97%
(±)-1-Phenyl-1,2-ethanediol
Formula: C₆H₅CH(OH)CH₂OH
MW: 138.17 g/mol
Boiling Pt: 272…274 °C (1013 hPa)
Melting Pt: 61…64 °C
MDL Number: MFCD00003546
CAS Number: 93-56-1
EINECS: 202-258-1
Merck Index: 12,09029

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR