Sodium pyruvate ≥99%, ReagentPlus®

Synonyms: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt

P2256-25G P2256-5G P2256-100G P2256-500G P2256-10MG
220200-350EA 41.77 USD
220200-350 220200-352 220200-348 220200-351 220200-349
Sodium pyruvate ≥99%, ReagentPlus®
Sodium pyruvate
Formula: CH₃COCOONa
MW: 110.04 g/mol
Melting Pt: 220…230 °C
Density: 1.718 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002586
CAS Number: 113-24-6
EINECS: 204-024-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR