4-Phenylbutyric acid ≥99%

Synonyms: 4-Phenylbutanoic acid

P21005-25G P21005-100G
220202-783EA 68.38 USD
220202-783 220202-782
4-Phenylbutyric acid ≥99%
4-Phenylbutyric acid
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 290 - 294 °C (1013 hPa)
Melting Pt: 48 - 51 °C
MDL Number: MFCD00004403
CAS Number: 1821-12-1
EINECS: 217-341-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR