Propionic acid ≥99%, Reagent Grade

Danger

Synonyms: Propanoic acid

P1386-1L P1386-500ML
220202-450EA 54.69 USD
220202-450 220202-451
Propionic acid ≥99%, Reagent Grade
Propionic acid
Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 141 °C (1013 hPa)
Melting Pt: -21 °C
Density: 0.99 g/cm³ (20 °C)
Flash Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00002756
CAS Number: 79-09-4
EINECS: 201-176-3
UN: 3463
ADR: 8,II
REACH: 01-2119486971-24
Merck Index: 13,07917

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR