(±)-1-Phenylethanol ≥98%

Danger

Synonyms: (±)-sec-Phenethyl alcohol, Styrene alcohol, (±)-α-Methylbenzyl alcohol, (±)-1-Phenylethyl alcohol, 1-Phenylethanol, Methyl phenyl carbinol, α-Methylbenzyl alcohol, Styrallyl alcohol

P13800-25G P13800-500G P13800-1KG
220202-445EA 38.23 USD
220202-445 220202-446 220202-444
(±)-1-Phenylethanol ≥98%
(±)-1-Phenylethanol
Formula: C₆H₅CH(OH)CH₃
MW: 122.17 g/mol
Boiling Pt: 401 °C (1013 hPa)
Melting Pt: 20 °C
Density: 1.015 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004508
CAS Number: 98-85-1
EINECS: 202-707-1
UN: 2937
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa955c5829772d9b27ccf96aa5e9b66b661e94a5&vid=43f0c1e8ef3c916809a95af92edf59ec33769eab&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR