L(-)-Proline, ReagentPlus®

Synonyms: (S)-(-)-ProlineH-Pro-OHL-Proline, H-Pro-OH

P0380-100G P0380-10MG P0380-5KG P0380-1KG
220202-055EA 125.54 USD
220202-055 220202-056 220202-058 220202-057
L(-)-Proline, ReagentPlus®
L(-)-Proline
Formula: C₅H₉NO₂
MW: 115.13 g/mol
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 220…222 °C
Density: 1.35 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064318
CAS Number: 147-85-3
EINECS: 205-702-2
Merck Index: 13,07871

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR