Oxalyl dichloride ≥98%, Reagent Grade

Danger

Synonyms: Oxalyl chloride

O8801-2.5KG O8801-1KG O8801-100G O8801-25G
220201-772EA 512.31 USD
220201-772 220201-771 220201-770 220201-773
Oxalyl dichloride ≥98%, Reagent Grade
Oxalyl dichloride
Formula: C₂Cl₂O₂
MW: 126.93 g/mol
Boiling Pt: 63 °C (1013 hPa)
Melting Pt: –12 °C
Density: 1.48 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00000704
CAS Number: 79-37-8
EINECS: 201-200-2
UN: 3129
ADR: 4.3,I
Merck Index: 12,07046

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR