Oleyl amine ≥70% (by GC), Technical Grade

Synonyms: cis-9-Octadecenylamine, (9Z)-9-Octadecen-1-amine, (Z)-Octadec-9-en-1-amine

O7805-500G O7805-100G O7805-5G
220201-376EA 224.92 USD
220201-376 220201-375 220201-377
Oleyl amine ≥70% (by GC), Technical Grade
Oleyl amine
Formula: C₁₈H₃₇N
MW: 267.5 g/mol
Boiling Pt: 364 °C (1013 hPa)
Melting Pt: 21 °C
Density: 0.813 g/cm³ (20 °C)
MDL Number: MFCD00066507
CAS Number: 112-90-3
EINECS: 204-015-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR