Sodium oleate

Synonyms: Oleic acid sodium salt, 9Z-Octadecenoic Acid, C18:1(9Z), cis-9-Octadecenoic Acid

O7501-1G O7501-10G O7501-5G O7501-250MG
220200-211EA 171.08 USD
220200-211 220200-210 220200-213 220200-212
Sodium oleate
Sodium oleate
Formula: C₁₈H₃₃NaO₂
MW: 304.45 g/mol
Melting Pt: 232…235 °C
Density: 1.02 g/cm³ (20 °C)
Storage Temperature: Freezer
MDL Number: MFCD00004438
CAS Number: 143-19-1
EINECS: 205-591-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR