Oleic acid

Synonyms: (9Z)-9-Octadecenoic acid

O1383-5G O1383-1G O1383-25G
220201-256EA 131.82 USD
220201-256 220201-254 220201-255
Oleic acid
Oleic acid
Formula: C₁₈H₃₄O₂
MW: 282.47 g/mol
Boiling Pt: 286 °C (1013 hPa)
Melting Pt: 16.7 °C
Density: 0.8946 g/cm³ (20 °C)
Flash Pt: 180 °C
Storage Temperature: Freezer
MDL Number: MFCD00064242
CAS Number: 112-80-1
EINECS: 204-007-1
Merck Index: 13,06898
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5aecc5dce4d0318386a45d7adb7c5822631010a&vid=892a2df4ecab3679b06983b84cd27ad334ef961f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR