Nitro blue tetrazolium chloride (NBT) for microscopy

Warning

Synonyms: 5,5'-Diphenyl-3,3'-bis(4-nitrophenyl)-2,2'-(3,3'-dimethoxybiphenyl-4,4'-ylene)ditetrazolium dichloride

N6639-250MG N6639-50MG N6639-1G
220200-721EA 140.31 USD
220200-721 220200-722 220200-720
Nitro blue tetrazolium chloride (NBT) for microscopy
Nitro blue tetrazolium chloride (NBT)
Formula: C₄₀H₃₀Cl₂N₁₀O₆
MW: 817.65 g/mol
Melting Pt: 200 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012159
CAS Number: 298-83-9
EINECS: 206-067-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR