Ninhydrin

Warning

N4876-100G N4876-500G
220200-525EA 144.62 USD
220200-525 220200-526
Ninhydrin
Ninhydrin
Formula: C₉H₆O₄
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: 250…258 °C
Density: 0.862 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003791
CAS Number: 485-47-2
EINECS: 207-618-1
Merck Index: 13,06582
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0ff1a5cd0c772262dac61a6c63dbdeca97baa2c&vid=61d6c7dbd1fe019587f026a878ce0951ce80fcc8&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR