Nitrobenzene ≥99%, ReagentPlus®

Danger

N10950-18L N10950-1L N10950-4L
220199-708EA 198.15 USD
220199-708 220199-709 220199-710
Nitrobenzene ≥99%, ReagentPlus®
Nitrobenzene
Formula: C₆H₅NO₂
MW: 123.11 g/mol
Boiling Pt: 210.8 °C (1013 hPa)
Melting Pt: 5.7 °C
Density: 1.2037 g/cm³ (20 °C)
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00007043
CAS Number: 98-95-3
EINECS: 202-716-0
UN: 1662
ADR: 6.1,II
Merck Index: 13,06621

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR