L-Methionine ≥98%, Reagent Grade

Synonyms: (S)-(-)-2-Amino-4-(methylthio)butyric acid, (S)-(-)-Methionine, L(-)-Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH, Methionine, (S)-2-Amino-4-(methylthio)butanoic acid

M9625-5G M9625-25G M9625-1KG M9625-100G M9625-500G
220199-414EA 19.2 USD
220199-414 220199-412 220199-411 220199-410 220199-413
L-Methionine ≥98%, Reagent Grade
L-Methionine
Formula: C₅H₁₁NO₂S
MW: 149.21 g/mol
Boiling Pt: 278 °C (1013 hPa)
Melting Pt: 136 °C
Density: 1.373 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00063097
CAS Number: 63-68-3
EINECS: 200-562-9
Merck Index: 13,06004

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR