Methyl-4-hydroxybenzoate analytical standard

Warning

Synonyms: Methylparaben, 4-Hydroxybenzoic acid methyl ester, Methyl Parahydroxybenzoate, Methyl 4-hydroxybenzoate

M8911-5VL M8911-1VL
220199-223EA 784.34 USD
220199-223 220199-222
Methyl-4-hydroxybenzoate analytical standard
Methyl-4-hydroxybenzoate
Formula: HOC₆H₄CO₂CH₃
MW: 152.15 g/mol
Boiling Pt: 270…280 °C (1013 hPa)
Melting Pt: 125…128 °C
Density: 1.1208 g/cm³ (137 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002352
CAS Number: 99-76-3
EINECS: 202-785-7
Merck Index: 13,06129
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84321052052f4be2a53d8d7ebac7fabe6fc5a435&vid=ede765fad275b2515a8c57ba4499bff313a18983&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR