Methyl salicylate ≥99%, ReagentPlus®

Warning

Synonyms: Wintergreen Oil, Methyl 2-hydroxybenzoate

M6752-1L M6752-250ML M6752-1GA M6752-500ML
220196-860EA 107.77 USD
220196-860 220196-861 220196-859 220196-862
Methyl salicylate ≥99%, ReagentPlus®
Methyl salicylate
Formula: 2-(HO)C₆H₄CO₂CH₃
MW: 152.15 g/mol
Boiling Pt: 432 °C (1013 hPa)
Melting Pt: 100…101 °C
Density: 1.184 g/cm³ (20 °C)
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00002214
CAS Number: 119-36-8
EINECS: 204-317-7
Merck Index: 13,06143
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d636eb895c055579d494c4b8ed1636c32f052dd7&vid=7f7da4bc95980831012b381705f4b5a3f4ab7003&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR