4-Methylbenzylamine ≥97%

Danger

Synonyms: 4-(Aminomethyl)toluene

M31201-100G M31201-25G
220197-522EA 291.85 USD
220197-522 220197-523
4-Methylbenzylamine ≥97%
4-Methylbenzylamine
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 195 °C (1013 hPa)
Melting Pt: 20…21 °C
Density: 0.952 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008123
CAS Number: 104-84-7
EINECS: 203-243-2
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR