Manganese(III) acetylacetonate, Technical Grade

Synonyms: Manganese (III) tris(2,4-pentanedionato-O,O'), Tris(2,4-pentanedionato-O,O')manganese (III), Manganese (III) 2,4-pentanedionate

M2284-25G M2284-100G
220196-806EA 45.69 USD
220196-806 220196-805
Manganese(III) acetylacetonate, Technical Grade
Manganese(III) acetylacetonate
Formula: C₁₅H₂₁MnO₆
MW: 352.27 g/mol
Melting Pt: 160 °C
MDL Number: MFCD00000023
CAS Number: 14284-89-0
EINECS: 238-188-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR