1-(2-Methoxyphenyl)piperazine

M22601-25G M22601-5G
220196-788EA 107.69 USD
220196-788 220196-789
1-(2-Methoxyphenyl)piperazine
1-(2-Methoxyphenyl)piperazine
Formula: C₁₁H₁₆N₂O
MW: 192.26 g/mol
Boiling Pt: 380 °C (1013 hPa)
Melting Pt: 35…38 °C
Density: 1.09 g/cm³ (20 °C)
MDL Number: MFCD00005958
CAS Number: 35386-24-4
EINECS: 252-537-7
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR