Magnesium sulfate heptahydrate ≥99.0%, ReagentPlus®

M1880-1KG M1880-5KG M1880-500G
220197-187EA 50.34 USD
220197-187 220197-189 220197-188
Magnesium sulfate heptahydrate ≥99.0%, ReagentPlus®
Magnesium sulphate heptahydrate
Formula: MgSO₄·7H₂O
Melting Pt: 1124 °C
Density: 2.66 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149785
CAS Number: 10034-99-8
EINECS: 231-298-2
Merck Index: 13,05715

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR