Malononitrile ≥99%

Danger

M1407-100G-A M1407-5G-A M1407-500G-A
220194-655EA 30.91 USD
220194-655 220194-657 220194-656
Malononitrile ≥99%
Malononitrile
Formula: C₃H₂N₂
MW: 66.06 g/mol
Boiling Pt: 220 °C (1013 hPa)
Melting Pt: 30 - 32 °C
Density: 1.191 g/cm³ (20 °C)
Flash Pt: 112 °C
MDL Number: MFCD00001883
CAS Number: 109-77-3
EINECS: 203-703-2
UN: 2647
ADR: 6.1,II
Merck Index: 13,05733

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR