Magnesium hydrogen orthophosphate trihydrate

M1265-1KG M1265-250G
220197-004EA 97.48 USD
220197-004 220197-005
Magnesium hydrogen orthophosphate trihydrate
Magnesium hydrogen orthophosphate trihydrate
Formula: HMgO₄P·3H₂O
MW: 174.33 g/mol
Density: 2.131 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150390
CAS Number: 7782-75-4
EINECS: 231-823-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR