Mercury(II) chloride, ReagentPlus®

Danger

Synonyms: Mercuric chloride, Mercury Chloride, Mercury dichloride

M1136-100G M1136-500G M1136-25G M1136-1KG
220196-966EA 48.8 USD
220196-966 220196-969 220196-968 220196-967
Mercury(II) chloride, ReagentPlus®
Mercury(II) chloride
Formula: HgCl₂
MW: 271.5 g/mol
Boiling Pt: 302 °C (1013 hPa)
Melting Pt: 273 °C
Density: 5.44 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011041
CAS Number: 7487-94-7
EINECS: 231-299-8
UN: 1624
ADR: 6.1 II
Merck Index: 13,05901

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR