Maleic acid, ReagentPlus®

Warning

M0375-500G M0375-25KG M0375-1KG M0375-100G
220196-531EA 39.77 USD
220196-531 220196-530 220196-529 220196-528
Maleic acid, ReagentPlus®
Maleic acid
Formula: C₄H₄O₄
MW: 116.07 g/mol
Boiling Pt: 256 °C (1013 hPa)
Melting Pt: 130…131 °C
Density: 1.59 g/cm³ (20 °C)
Flash Pt: 127 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00063177
CAS Number: 110-16-7
EINECS: 203-742-5
UN: 3261
ADR: 8,III
Merck Index: 13,05726
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3a1ea1f5bca5d5cf7fd40530f642f47db88908a&vid=1ac7f8cee2cd0433dbfb6e9271d41fe6ecc382c1&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR