L(+)-Leucine ≥98%, Reagent Grade

Synonyms: (S)-(+)-Leucine, Leu, H-Leu-OH, Leucine, L-Leucine

L8000-50G L8000-100G L8000-25G L8000-1KG L8000-250G
220196-049EA 76.18 USD
220196-049 220196-045 220196-048 220196-046 220196-047
L(+)-Leucine ≥98%, Reagent Grade
L(+)-Leucine
Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Melting Pt: 300 °C
Density: 1.165 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002617
CAS Number: 61-90-5
EINECS: 200-522-0
Merck Index: 14,05451

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR