L(+)-Lysine monohydrochloride ≥98%, Reagent Grade

L5626-1KG L5626-5KG L5626-100G L5626-500G
220194-716EA 189.54 USD
220194-716 220194-718 220194-715 220194-717
L(+)-Lysine monohydrochloride ≥98%, Reagent Grade
L(+)-Lysine monohydrochloride
Formula: H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
Melting Pt: 263 - 264 °C
Storage Temperature: Ambient
MDL Number: MFCD00064564
CAS Number: 657-27-2
EINECS: 211-519-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR