Lauric acid ≥98%

Warning

Synonyms: Dodecanoic acid, Dodecylic acid

L556-1KG L556-25G L556-3KG
220194-711EA 57.18 USD
220194-711 220194-712 220194-713
Lauric acid ≥98%
Lauric acid
Formula: C₁₂H₂₄O₂
MW: 200.32 g/mol
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 42…45 °C
Density: 0.875 g/cm³ (20 °C)
Flash Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00002736
CAS Number: 143-07-7
EINECS: 205-582-1
Merck Index: 13,05400

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR