3,5-Lutidine

Danger

Synonyms: 3,5-Dimethylpyridine

L4206-100ML L4206-500ML
220195-490EA 70 USD
220195-490 220195-491
3,5-Lutidine
3,5-Lutidine
Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 172.7 °C (1013 hPa)
Melting Pt: –9 °C
Density: 0.94 g/cm³ (20 °C)
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00006404
CAS Number: 591-22-0
EINECS: 209-708-6
UN: 1992
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR