2,6-Lutidine, ReagentPlus®

Synonyms: 2,6-Dimethylpyridine

L3900-2L-A L3900-500ML-A L3900-5G L3900-100ML-A
220195-473EA 241.54 USD
220195-473 220195-474 220195-475 220195-472
2,6-Lutidine, ReagentPlus®
2,6-Lutidine
Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 143.7 °C (1013 hPa)
Melting Pt: -6 °C
Density: 0.9252 g/cm³ (20 °C)
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00006345
CAS Number: 108-48-5
EINECS: 203-587-3
UN: 1993
ADR: 3,III
Merck Index: 13,05636

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR