L(+)-Isoleucine ≥98%, Reagent Grade

Synonyms: L-Isoleucine, (2S,3S)-(+)-Isoleucine, H-Ile-OH

I2752-100G I2752-1G I2752-10G I2752-25G I2752-1KG I2752-5G
220191-788EA 230.15 USD
220191-788 220192-683 220191-789 220192-685 220192-684 220192-686
L(+)-Isoleucine ≥98%, Reagent Grade
L(+)-Isoleucine
Formula: H₃CCH₂CH(CH₃)CH(NH₂)COOH
MW: 131.17 g/mol
Melting Pt: 279…280 °C
Storage Temperature: Ambient
MDL Number: MFCD00064222
CAS Number: 73-32-5
EINECS: 200-798-2
Merck Index: 14,05179

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR