Diisopropyl sulfide ≥99%

Synonyms: Isopropyl sulphide

I26809-100G I26809-25G
220191-785EA 371.54 USD
220191-785 220191-786
Diisopropyl sulfide ≥99%
Diisopropyl sulphide
Formula: C₆H₁₄S
MW: 118.24 g/mol
Boiling Pt: 67 °C (1013 hPa)
Melting Pt: –69.1 °C
Density: 0.8306 g/cm³ (0 °C)
MDL Number: MFCD00008895
CAS Number: 625-80-9
EINECS: 210-911-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR