1-Heptanesulfonic acid sodium salt

Synonyms: Sodium heptane-1-sulphonate, Sodium 1-heptanesulphonate

H8901-25G H8901-5G
220182-763EA 403.82 USD
220182-763 220182-764
1-Heptanesulfonic acid sodium salt
1-Heptanesulphonic acid sodium salt
Formula: H₃C(CH₂)₅CH₂SO₃Na
MW: 202.25 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007543
CAS Number: 22767-50-6
EINECS: 245-210-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95c09a5691140bab7446d88919f01593e8f0b9b2&vid=7c237cde5db01d0567ff19964ca3287434747480&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR